Anti-IRS1 Rabbit Polyclonal Antibody

Leverancier: Biorbyt
BIRBORB6241-100EA 306 EUR
BIRBORB6241-100 BIRBORB6241-20
Anti-IRS1 Rabbit Polyclonal Antibody
Antilichamen
Western Blot: 1:500-1:2000; Immunofluorescence: 1:100-1:500

Type: Primary
Antigen: IRS1 (insulin receptor substrate 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR