Anti-CERS1 Rabbit polyclonal antibody
Leverancier: Biorbyt
BIRBORB6296-100EA 0 EUR
BIRBORB6296-100 BIRBORB6296-200
Anti-CERS1 Rabbit polyclonal antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: CERS1 (ceramide synthase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat