Anti-CRTC2 Rabbit polyclonal antibody
Leverancier: Biorbyt
BIRBORB6370-100EA 0 EUR
BIRBORB6370-100 BIRBORB6370-200
Anti-CRTC2 Rabbit polyclonal antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: CRTC2 (CREB regulated transcription coactivator 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestel nu