Anti-MYL1 Rabbit Polyclonal Antibody
Leverancier: Biorbyt
BIRBORB6426-100EA 0 EUR
BIRBORB6426-100 BIRBORB6426-200
Anti-MYL1 Rabbit Polyclonal Antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: MYL1 (myosin, light chain 1, alkali; skeletal, fast)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestel nu