Anti-MYT1 Rabbit Polyclonal Antibody
Leverancier: Biorbyt
BIRBORB6431-100EA 0 EUR
BIRBORB6431-100 BIRBORB6431-200
Anti-MYT1 Rabbit Polyclonal Antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: MYT1 (myelin transcription factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestel nu