Anti-RHEB Rabbit Polyclonal Antibody
Leverancier: Biorbyt
BIRBORB6435-200EA 0 EUR
BIRBORB6435-200 BIRBORB6435-100
Anti-RHEB Rabbit Polyclonal Antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500
Type: Primary
Antigen: RHEB (Ras homolog enriched in brain)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat