Anti-NFKB2 Rabbit Polyclonal Antibody
Leverancier: Biorbyt
BIRBORB6500-200EA 0 EUR
BIRBORB6500-200 BIRBORB6500-100
Anti-NFKB2 Rabbit Polyclonal Antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: NFKB2 (nuclear factor of kappa light polypeptide gene enhancer in B-cells 2 (p49/p100))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestel nu