Anti-CDKN1A Rabbit polyclonal antibody

Leverancier: Biorbyt
BIRBORB6568-5EA 51 EUR
BIRBORB6568-5 BIRBORB6568-20 BIRBORB6568-100
Anti-CDKN1A Rabbit polyclonal antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: CDKN1A (cyclin-dependent kinase inhibitor 1A (p21, Cip1))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR