Anti-PAK1/2/3 Rabbit Polyclonal Antibody
Leverancier: Biorbyt
BIRBORB6636-200EA 0 EUR
BIRBORB6636-200 BIRBORB6636-100
Anti-PAK1/2/3 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: PAK1/2/3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat