Anti-RGS19 G Rabbit Polyclonal Antibody
Leverancier: Biorbyt
BIRBORB6874-100EA 0 EUR
BIRBORB6874-100 BIRBORB6874-200
Anti-RGS19 G Rabbit Polyclonal Antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500
Type: Primary
Antigen: RGS19 G
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat