Anti-S100A8 Rabbit Polyclonal Antibody

Leverancier: Biorbyt
BIRBORB6912-200EA 508 EUR
BIRBORB6912-200 BIRBORB6912-100
Anti-S100A8 Rabbit Polyclonal Antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500

Type: Primary
Antigen: S100A8 (S100 calcium binding protein A8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR