Anti-SLC25A11 Rabbit Polyclonal Antibody

Leverancier: Biorbyt
BIRBORB6966-100EA 312 EUR
BIRBORB6966-100 BIRBORB6966-200
Anti-SLC25A11 Rabbit Polyclonal Antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: SLC25A11 (solute carrier family 25 (mitochondrial carrier; oxoglutarate carrier), member 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR