Anti-STAT5A Rabbit Polyclonal Antibody

Leverancier: Biorbyt
BIRBORB7021-100EA 0 EUR
BIRBORB7021-100
Anti-STAT5A Rabbit Polyclonal Antibody
Antilichamen
  • Western Blot: 1:500-1:1000 Immunochemistry: 1:50-1:100

Type: Primary
Antigen: STAT5A (signal transducer and activator of transcription 5A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR