Anti-WNT10B Rabbit Polyclonal Antibody

Leverancier: Biorbyt
BIRBORB7140-200EA 508 EUR
BIRBORB7140-200 BIRBORB7140-100
Anti-WNT10B Rabbit Polyclonal Antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: WNT10B (wingless-type MMTV integration site family, member 10B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR