Anti-TRIM39 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Leverancier: Biorbyt
BIRBORB7206-100EA 0 EUR
BIRBORB7206-100
Anti-TRIM39 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antilichamen
Type: Primary
Antigen: TRIM39 (tripartite motif containing 39)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR