Anti-RNF24 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Leverancier: Biorbyt
BIRBORB7207-100EA 385 EUR
BIRBORB7207-100
Anti-RNF24 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antilichamen
Type: Primary
Antigen: RNF24 (ring finger protein 24)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR