Anti-C1orf74 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Leverancier: Biorbyt
BIRBORB7448-100EA 385 EUR
BIRBORB7448-100
Anti-C1orf74 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antilichamen
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: C1orf74 (chromosome 1 open reading frame 74)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR