Anti-RP1 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Leverancier: Biorbyt
BIRBORB7823-100EA 385 EUR
BIRBORB7823-100
Anti-RP1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antilichamen
Type: Primary
Antigen: RP1 (retinitis pigmentosa 1 (autosomal dominant))
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR