Anti-CD2 Mouse Monoclonal Antibody

Leverancier: Biorbyt
BIRBORB8087-100EA 1195 EUR
BIRBORB8087-100
Anti-CD2 Mouse Monoclonal Antibody
Antilichamen
Western Blot: 1:1000-1:3000

Type: Primary
Antigen: CD2 (CD2 molecule)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a (Mouse)
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR