Anti-FBL Rabbit Polyclonal Antibody (FITC (Fluorescein))
Leverancier: Biorbyt
BIRBORB8096-100EA 0 EUR
BIRBORB8096-100
Anti-FBL Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antilichamen
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: FBL (fibrillarin)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestel nu