Anti-PTPRC Mouse Monoclonal Antibody (PE (Phycoerythrin))

Leverancier: Biorbyt
BIRBORB8187-100EA 667 EUR
BIRBORB8187-100
Anti-PTPRC Mouse Monoclonal Antibody (PE (Phycoerythrin))
Antilichamen
Western Blot: 1:1000-1:3000

Type: Primary
Antigen: PTPRC (protein tyrosine phosphatase, receptor type, C)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a (Mouse)
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR