Anti-FAS Mouse Monoclonal Antibody (PE (Phycoerythrin))
Leverancier: Biorbyt
BIRBORB8219-100EA 0 EUR
BIRBORB8219-100
Anti-FAS Mouse Monoclonal Antibody (PE (Phycoerythrin))
Antilichamen
Western Blot: 1:1000-1:3000
Type: Primary
Antigen: FAS (Fas cell surface death receptor)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human