Anti-MAP3K5 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Leverancier: Biorbyt
BIRBORB8575-100EA 0 EUR
BIRBORB8575-100
Anti-MAP3K5 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antilichamen
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: MAP3K5 (mitogen-activated protein kinase kinase kinase 5)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestel nu