Anti-HIRA Rabbit Polyclonal Antibody (FITC (Fluorescein))

Leverancier: Biorbyt
BIRBORB8998-100EA 385 EUR
BIRBORB8998-100
Anti-HIRA Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antilichamen
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: HIRA (histone cell cycle regulator)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR