Anti-SULT2A1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Leverancier: Biorbyt
BIRBORB9357-100EA 0 EUR
BIRBORB9357-100
Anti-SULT2A1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antilichamen
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: SULT2A1 (sulfotransferase family, cytosolic, 2A, dehydroepiandrosterone (DHEA)-preferring, member 1)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat