Anti-PSAP Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverancier: Biorbyt
BIRBORB9657-100EA 385 EUR
BIRBORB9657-100
Anti-PSAP Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antilichamen
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: PSAP
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR