Anti-SLY 2 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Leverancier: Biorbyt
BIRBORB9877-100EA 0 EUR
BIRBORB9877-100
Anti-SLY 2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antilichamen
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: SLY 2
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR