Anti-ATG4B Rabbit polyclonal antibody
Leverancier: Biorbyt
BIRBORB10113-200EA 0 EUR
BIRBORB10113-200 BIRBORB10113-100
Anti-ATG4B Rabbit polyclonal antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: ATG4B (autophagy related 4B, cysteine peptidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestel nu