Anti-BRCA1 Rabbit polyclonal antibody
Leverancier: Biorbyt
BIRBORB10202-100EA 0 EUR
BIRBORB10202-100 BIRBORB10202-200
Anti-BRCA1 Rabbit polyclonal antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: BRCA1 (breast cancer 1, early onset)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat