Anti-BRMS1 Rabbit polyclonal antibody
Leverancier: Biorbyt
BIRBORB10210-200EA 0 EUR
BIRBORB10210-200 BIRBORB10210-100
Anti-BRMS1 Rabbit polyclonal antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: BRMS1 (breast cancer metastasis suppressor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestel nu