Anti-CAPN1 Rabbit Polyclonal Antibody

Leverancier: Biorbyt
BIRBORB10219-200EA 508 EUR
BIRBORB10219-200 BIRBORB10219-100
Anti-CAPN1 Rabbit Polyclonal Antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: CAPN1 (calpain 1, (mu/I) large subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR