Anti-CCL21 Rabbit polyclonal antibody

Leverancier: Biorbyt
BIRBORB10267-200EA 508 EUR
BIRBORB10267-200 BIRBORB10267-100
Anti-CCL21 Rabbit polyclonal antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: CCL21 (chemokine (C-C motif) ligand 21)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR