Anti-CCR6 Rabbit Polyclonal Antibody

Leverancier: Biorbyt
BIRBORB10274-20EA 109 EUR
BIRBORB10274-20 BIRBORB10274-100 BIRBORB10274-200 BIRBORB10274-5
Anti-CCR6 Rabbit Polyclonal Antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: CCR6 (chemokine (C-C motif) receptor 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR