Anti-CXCR6 Rabbit Polyclonal Antibody
Leverancier: Biorbyt
BIRBORB10275-100EA 0 EUR
BIRBORB10275-100 BIRBORB10275-200
Anti-CXCR6 Rabbit Polyclonal Antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: CXCR6 (chemokine (C-X-C motif) receptor 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat