Anti-DNMT3B Rabbit Polyclonal Antibody
Leverancier: Biorbyt
BIRBORB10548-200EA 0 EUR
BIRBORB10548-200 BIRBORB10548-100
Anti-DNMT3B Rabbit Polyclonal Antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: DNMT3B (DNA (cytosine-5-)-methyltransferase 3 beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat