Anti-Duck viral hepatitis Rabbit Polyclonal Antibody

Leverancier: Biorbyt
BIRBORB10557-200EA 522 EUR
BIRBORB10557-200 BIRBORB10557-100
Anti-Duck viral hepatitis Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: Duck viral hepatitis
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Duck
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR