Anti-Brucella Rabbit polyclonal antibody

Leverancier: Biorbyt
BIRBORB10564-100EA 0 EUR
BIRBORB10564-100 BIRBORB10564-200
Anti-Brucella Rabbit polyclonal antibody
Antilichamen
Type: Primary
Antigen: Brucella
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR