Anti-F8 Rabbit polyclonal antibody
Leverancier: Biorbyt
BIRBORB10633-200EA 0 EUR
BIRBORB10633-200 BIRBORB10633-100
Anti-F8 Rabbit polyclonal antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: F8 (coagulation factor VIII, procoagulant component)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestel nu