Anti-APOA1 Rabbit Polyclonal Antibody

Leverancier: Biorbyt
BIRBORB10643-200EA 508 EUR
BIRBORB10643-200 BIRBORB10643-100
Anti-APOA1 Rabbit Polyclonal Antibody
Antilichamen
  • Western Blot: 1:100-500

Type: Primary
Antigen: APOA1 (apolipoprotein A-I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR