Anti-FGF3 Rabbit Polyclonal Antibody
Leverancier: Biorbyt
BIRBORB10649-200EA 0 EUR
BIRBORB10649-200 BIRBORB10649-100
Anti-FGF3 Rabbit Polyclonal Antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: FGF3 (fibroblast growth factor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat