Anti-GAPDH Mouse monoclonal antibody

Leverancier: Biorbyt
BIRBORB10693-100EA 312 EUR
BIRBORB10693-100 BIRBORB10693-20 BIRBORB10693-200 BIRBORB10693-5
Anti-GAPDH Mouse monoclonal antibody
Antilichamen
  • ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: GAPDH (glyceraldehyde-3-phosphate dehydrogenase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR