Anti-GHRL Rabbit Polyclonal Antibody
Leverancier: Biorbyt
BIRBORB10713-20EA 0 EUR
BIRBORB10713-20 BIRBORB10713-5 BIRBORB10713-100 BIRBORB10713-200
Anti-GHRL Rabbit Polyclonal Antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: GHRL (ghrelin/obestatin prepropeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestel nu