Anti-GCG Mouse monoclonal antibody
Leverancier: Biorbyt
BIRBORB10721-100EA 0 EUR
BIRBORB10721-100 BIRBORB10721-200
Anti-GCG Mouse monoclonal antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: GCG (glucagon)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat