Anti-Glycine Receptor alpha 1 Rabbit polyclonal antibody

Leverancier: Biorbyt
BIRBORB10729-200EA 508 EUR
BIRBORB10729-200 BIRBORB10729-100
Anti-Glycine Receptor alpha 1 Rabbit polyclonal antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: Glycine Receptor alpha 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR