Anti-GPA33 Rabbit Polyclonal Antibody
Leverancier: Biorbyt
BIRBORB10734-100EA 312 EUR
BIRBORB10734-100 BIRBORB10734-200
Anti-GPA33 Rabbit Polyclonal Antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: GPA33 (glycoprotein A33 (transmembrane))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat