Anti-HBsAg Goat polyclonal antibody
Leverancier: Biorbyt
BIRBORB10771-200EA 0 EUR
BIRBORB10771-200 BIRBORB10771-100
Anti-HBsAg Goat polyclonal antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: HBsAg
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Virus