Anti-HbeAg Mouse monoclonal antibody

Leverancier: Biorbyt
BIRBORB10776-20EA 109 EUR
BIRBORB10776-20 BIRBORB10776-100 BIRBORB10776-200 BIRBORB10776-1 BIRBORB10776-5
Anti-HbeAg Mouse monoclonal antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: HbeAg
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Virus
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR