Anti-HSP90 beta Rabbit Polyclonal Antibody
Leverancier: Biorbyt
BIRBORB10852-100EA 0 EUR
BIRBORB10852-100 BIRBORB10852-5 BIRBORB10852-200 BIRBORB10852-20
Anti-HSP90 beta Rabbit Polyclonal Antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: HSP90 beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestel nu