Anti-IgM µ Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverancier: Rockland Immunochemicals
ROCK211-1207EA 830 EUR
ROCK211-1207
Anti-IgM µ Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antilichamen
Secondary Goat Anti-IgM mu chain Reacts with Rabbit (Lapine)

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 2.0 ml

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: µ-chain
Host: Goat
Isotype: IgM
Reactivity: Rabbit
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR