Anti-IgG F(ab')2 Rabbit Polyclonal Antibody (Biotin)
Leverancier: Rockland Immunochemicals
ROCK612-4604EA 382 EUR
ROCK612-4604
Anti-IgG F(ab')2 Rabbit Polyclonal Antibody (Biotin)
Antilichamen
Secondary Rabbit Anti-F(ab')2 Reacts with Rat
Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: F(ab')2
Host: Rabbit
Isotype: IgG
Reactivity: Rat